missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLOBINclear™ Kit, mouse/rat, for globin mRNA depletion

Product Code. 10237977
Change view
Click to view available options
Quantity:
20 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10237977 20 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10237977 Supplier Invitrogen™ Supplier No. AM1981
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

20μL Capture Oligo Mix; 600μL Streptavidin Magnetic Beads; 1.5mL 2X Hybridization Buffer; 1.5mL Streptavidin Bead Buffer; 1.75 Nuclease-free Water; 200μL RNA Binding Beads; 5mL RNA Wash Solution Concentrate; 1mL Elution Buffer; 80μL RNA Bead Buffer; 4mL RNA Binding Buffer Concentrate; 100 1.5-ml Non-stick Tubes

Requires

Kit requires use of a magnetic stand. The 6 Tube Magnetic Stand (Cat. No. AM10055) can hold six separate 1.5mL conical microcentrifuge tubes or 2mL round-bottomed microtubes with a diameter of 8 to 10 mm.

Employs a novel, non-enzymatic technology that rapidly depletes >95% of the alpha and beta globin mRNA from total RNA preparations derived from whole blood

The Ambion™ GLOBINclear™-Mouse/Rat Kit employs a novel, non-enzymatic technology that rapidly depletes >95% of the alpha and beta globin mRNA from total RNA preparations derived from whole blood. The kit includes sufficient reagents for 20 reactions.

  • Remove >95% of unwanted globin mRNA from whole blood total RNA
  • Detect up to 50% more genes—analyze previously undetected genes
  • Achieve greater representation of the genome, low 3'/5' ratios
  • Avoid RNase H treatments that can degrade mRNA and alter expression profiles

It is widely known that expression array data generated from whole blood total RNA samples have reduced detection sensitivity compared to data from fractionated blood samples. Examination of whole blood total RNA reveals a dominant band in the 600–700 base region. This band represents globin mRNA, which is expressed at high levels in red blood cells (RBCs) and reticulocytes (RBC precursors). Up to 70% of the mRNA (by mass) in whole blood total RNA are globin transcripts. Array analysis using total RNA containing a high percentage of globin mRNA has been shown to:

  • Decrease present calls
  • Decrease call concordance
  • Increase signal variation

The GLOBINclear™ kit employs a novel hybridization technology that takes advantage of the strength of biotin/streptavidin binding, the specificity of nucleic acid hybridization, and the convenience of magnetic separations. There are no harsh RNase H enzymatic treatments that may compromise the expression profile by degrading mRNAs other than globin mRNA. The resulting mRNA is a superior template for synthesizing labeled cDNA for array analysis and is ideal for quantitative RT-PCR. The enrichment delivers a significant increase in Percent Present calls, decreased sample-to-sample variability, and low 3'/5' ratios.

Accessory Product:
The kit requires the use of a magnetic stand. The 6 Tube Magnetic Stand (SKU# AM10055) can hold six separate 1.5 mL conical microcentrifuge tubes or 2 mL round-bottomed microtubes with a diameter of 8–10 mm.

Order Info

Shipping Condition: Wet/Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Mouse/Rat Kit
Content And Storage • 20 μl Capture Oligo Mix (-20°C)
• 600 μl Streptavidin Magnetic Beads (4°C—do not freeze)
• 1.5 ml 2X Hybridization Buffer (room temperature)
• 1.5 ml Streptavidin Bead Buffer (room temperature)
• 1.75 Nuclease-free Water (-20°C, 4°C, or room temperature)
• 200 μl RNA Binding Beads (4°C—do not freeze)
• 5 ml RNA Wash Solution Concentrate (4°C or room temperature)
• 1 ml Elution Buffer (4°C or room temperature)
• 80 μl RNA Bead Buffer (room temperature)
• 4 ml RNA Binding Buffer Concentrate (room temperature)
• 100 1.5-ml Non-stick Tubes (room temperature)
Final Product Type Total RNA (Globin mRNA-Depleted)
High-throughput Compatibility High-throughput Compatible
Sample Type Total RNA (Whole Blood)
Species Mouse, Rat
Format Microtube
Product Line Ambion, GLOBINclear
For Use With (Application) Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting, cDNA Library Construction
Quantity 20 Preps

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.