missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GlycoBlue™ Coprecipitant (15 mg/mL)

Product Code. 10301575
Change view
Click to view available options
Quantity:
300 μL
5 x 300 μL
Unit Size:
1500µL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10301575 5 x 300 μL 1500µL
10391565 300 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10301575 Supplier Invitrogen™ Supplier No. AM9516
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant.

GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant. The attached dye increases visibility of the pellet. This product is an ideal coprecipitant in nuclease protection assays at 1/100 dilution of stock solution.

Features of GlycoBlue™ Coprecipitant:
• Ideal for RT-PCR
• Increases pellet mass and visibility
• Quantitative recovery of low concentrations (ng/mL) of nucleic acid
• Prevents pellet loss in nuclease protection assays

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 15 mg/mL
Content And Storage Store at –20°C.
Packaging Type Tube
Product Line Ambion, GlycoBlue
Purity Molecular Biology Grade
Quantity 5 x 300 μL
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.