missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mirVana™ miRNA Isolation Kit, with phenol

Product Code. 10763147
Change view
Click to view available options
Quantity:
40 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10763147 40 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10763147 Supplier Invitrogen™ Supplier No. AM1560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The mirVana™ miRNA Isolation Kit uses a rapid procedure to isolate small RNAs from tissue and cells using an efficient glass fiber filter (GFF)-based method.

The mirVana™ miRNA Isolation Kit uses a rapid procedure to isolate small RNAs from tissue and cells using an efficient glass fiber filter (GFF)-based method. The method isolates total RNA ranging in size from kilobases down to 10-mers. Each kit contains sufficient reagents and consumables for either 40 isolations of total RNA (including small RNAs), or 20 separate large and small RNA fractions.

Efficient recovery of small RNA
Current RNA purification procedures rely on organic extraction, followed by alcohol precipitation or adsorption of nucleic acid molecules on a GFF or silicate matrix. Unfortunately, the former procedure requires alcohol precipitation, which is time consuming and inefficiently recovers small RNAs. The latter procedure typically results in the loss of substantial amounts of small RNA. The mirVana miRNA Isolation Kit uses organic extraction followed by purification on a GFF under specialized binding and wash conditions. As a result, the kit effectively recovers all RNA—from large mRNA and ribosomal RNA down to 10-mers—from virtually all cell and tissue types. The mirVana miRNA Isolation Kit enables the isolation of small RNA-containing total RNA from samples consisting of 102–107 cultured cells or 0.5–250 mg tissue.

Enrich for small RNA
Although the mirVana miRNA Isolation Kit efficiently purifies all RNA larger than 10nt, it also includes procedures for isolating RNA fractions, specifically enriched or depleted in small RNA species. Through the enrichment procedure included in the mirVana miRNA Isolation Kit, RNA molecules of ∽200 nt and less can be efficiently purified away from the larger RNA species. The resulting RNA preparation is highly enriched for miRNAs, siRNAs, and/or snRNAs. Small RNA enrichment using the mirVana miRNA Isolation Kit procedure allows more sensitive small RNA detection with less background as compared to the same assay used with total RNA.

• Efficient isolation of small RNA-containing total RNA
• Enrich for small RNA (less than 200nt) to increase sensitivity in downstream analyses
• Simple 30 minute procedure
• Ideal for miRNA, siRNA, shRNA, and snRNA analysis
• Compatible with virtually all cell and tissue types

Accessory products
For tissues that have very high levels of ribonucleases, such as pancreas or spleen, the mirVana miRNA Isolation Kit is recommended, although it is not designed to recover protein and small RNA from the same sample.

Not high-throughput compatible (Manual)

TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage • 30 mL miRNA Wash Solution I (room temperature)
• 50 mL Wash Solution 2/3 (room temperature)
• 80 Collection Tubes (room temperature)
• 40 Filter Cartridges (room temperature)
• 100 mL Lysis/Binding Buffer (4°C)
• 10 mL miRNA Homogenate Additive (4°C)
• 100 mL Acid-Phenol:Chloroform (4°C)
• 1.4 mL Gel Loading Buffer II (-20°C)
• 5 mL Elution Solution (-20°C, 4°C, or room temperature)
Isolation Technology Spin Column (Glass Fiber Filter), Organic Extraction
Purification Time 30 min. (After sample homogenization)
Elution Volume 100 μL
Sample Type Bacteria, Cells, Plant, Tissue, Viruses, Yeast, Enzymatic Reactions
Final Product Type Small RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) qRT-PCR, microarray analysis, Northern blotting
Quantity 40 Preps
Shipping Condition Room Temperature
Starting Material Amount Bacteria: ≤107 cells
Cells: ≤107
Plant: ≤250 mg
Tissue: ≤250 mg
Virus: ≤107 cells
Yeast: ≤107 cells
Enzymatic Reactions: varies
Yield Cells: ≤16 μg RNA per 50,000 cells
Tissue: ≤160 μg per 5 mg
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • mirVana miRNA Isolation Kit, with phenol
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H301-Toxic if swallowed.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H412-Harmful to aquatic life with long lasting effects.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P330-Rinse mouth.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contains : phenol, chloroform,2-Mercaptoethanol, Thiocyanic acid, Formamide

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.