missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IPTG (Dioxane-Free)

Product Code. 10556735
Change view
Click to view available options
Quantity:
2 g
5 g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10556735 2 g Each
10685985 5 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10556735 Supplier Invitrogen™ Supplier No. AM9462

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage.

Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage. IPTG is also used to induce the expression of cloned genes that are under control of the lac operon.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material IPTG (Isopropyl β-D-thiogalactopyranoside)
Content And Storage Store at –20°C.
Packaging Type Bottle
Product Line Ambion
Quantity 2 g
Shipping Condition Room Temperature
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.