missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IPTG (Dioxane-Free)

Product Code. 10685985
Change view
Click to view available options
Quantity:
2 g
5 g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10685985 5 g Each
10556735 2 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10685985 Proveedor Invitrogen™ N.º de proveedor AM9464
Black Friday Starts Here
Use promo code 27984
Ver todas las promociones
 Restricted Item
In Stock
Añadir a la cesta
Edge
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage.

Invitrogen IPTG (Isopropyl-beta-D-thiogalactoside) is an inducer of the lac operon and is frequently used in conjunction with X-GAL to determine the lac phenotype of recombinant plasmids or phage. IPTG is also used to induce the expression of cloned genes that are under control of the lac operon.

Información de órdenes

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material IPTG (Isopropyl β-D-thiogalactopyranoside)
Content And Storage Store at –20°C.
Packaging Type Bottle
Product Line Ambion
Quantity 5 g
Shipping Condition Room Temperature
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.