missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagMAX™-96 DNA Multi-Sample Kit

Product Code. 12013729
Click to view available options
No. of Reactions:
480 Preps
96 Preps
Unit Size:
96 preps
Each
This item is not returnable. View return policy

Product Code. 12013729

Brand: Invitrogen™ 4413021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • DNA Binding Beads, 10 mg/mL, 1 × 1.8mL (Same for both kits) Store at 2 to 8 °C
  • DNA Elution Buffer 1 1 x 15mL (Same for both kits) Store at RT
  • DNA Elution Buffer 2 1 x 15mL (Same for both kits)Store at RT
  • Elution Plate, 96-well, 2 plates - Store at RT
  • Wash Solution 1 Concentrate, 1 x 15mL for PN 4413021 - Store at RT
  • Wash Solution 2 Concentrate, 1 x 50mL (Same for both kits) Store at RT
  • Multi-Sample DNA Lysis Buffer, 1 x 55mL (Same for both kits) Store at RT
  • MicroAmp Clear Adhesive Film, 1 x 4 films - Store at RT
  • Nuclease-Free Water 1 x 50mL (Same for both kits) Store at RT
  • PK Buffer 1 x 11mL (Same for both kits) Store at RT
  • Processing Plate, 96-well, 1 plate - Store at RT
  • Proteinase K, 1 x 850 μL - Store at -15 to -25 °C
  • Ribonuclease A (RNase A), 1 x 530 μL - Store at -15 to -25 °C

MagMAX™ DNA Multi-Sample 96-Well Kits are designed for rapid purification of high-quality genomic DNA from a variety of sample types including blood and blood cards, tissue, mouse tails, buccal swabs, buffy coats, and cultured cells.

MagMAX™ DNA Multi-Sample 96-Well Kits are designed for rapid purification of high-quality genomic DNA from a variety of sample types including blood and blood cards, tissue, mouse tails, buccal swabs, buffy coats, and cultured cells. These purification kits use MagMAX™ magnetic bead-based nucleic acid isolation technology to produce high yields of purified DNA, free from inhibitors that may affect downstream reactions such as genotyping assays, CE and next-generation sequencing, and high resolution melting analysis. The 96-prep and 5 x 96-prep kits are designed for higher throughput and automated format processing.

The demands for DNA yield and purity have become more stringent with the development of new analytical methods and technologies, such as high-throughput genotyping, and next-generation DNA sequencing platforms. MagMAX™ DNA Multi-Sample Kits meet the challenge by delivering highly pure DNA that is free of nucleases, proteins, and other inhibitors of downstream enzymatic reactions.

As shown in Table 1, DNA with high yield and purity can be obtained from a variety of materials using the MagMAX™ DNA Multi-Sample Kits. This is in contrast to filter membrane–based methods that often require different kit configurations for various input materials. For applications requiring maximal yields, for small batch sizes, or for DNA archival purposes MagMAX™ DNA Multi-Sample Kit (tube format, manual processing) may be desirable because larger sample inputs are accommodated. DNA yields suitable for most routine applications may be obtained using the MagMAX™-96 DNA Multi-Sample Kits (96-well plate format, compatible with automated handling) (Table 1).

The MagMAX™ DNA Multi-Sample Kit also delivers high yields of DNA from lipid or glycogen rich samples, such as brain and liver tissues, respectively. Figure 1 shows more than twice as much DNA was purified from tissues with the MagMAX™ DNA Multi-Sample Kit than with the leading competitor’s filter membrane–based method.

Most enzymatic molecular applications are prone to inhibition by analytes carried over from the sample matrix (e.g., heme, heparin, salts, protein) or the purification method itself (phenol, salts, alcohol). Figure 2 illustrates the effect of DNA purity on sensitive molecular applications, such as quantitative PCR (qPCR). DNA was extracted from blood using the MagMAX™-96 DNA Multi-Sample Kit, or a competitor’s filter membrane–based kit and subjected to real-time PCR analysis. The cycle threshold CT values obtained with the DNA purified using the MagMAX™ DNA Multi-Sample Kit were lower than those obtained with the filter membrane kit–purified DNA, indicating the MagMAX™ kit yields higher-purity DNA.

MagMAX™ DNA Multi-Sample Kits provide purified DNA that is suitable for a variety of Applied Biosystems application workflows (Figure 3), including SNP genotyping and DNA sequencing analysis (see Figure 4). To see the full performance description of DNA purified using the MagMAX™ DNA Multi-Sample Kits in TaqMan™ SNP genotyping assays and with BigDye™ chemistry in DNA sequence analysis, view the MagMAX™ DNA Multi-Sample Kit Product Bulletin.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
• DNA Binding Beads, 10 mg/mL, 1 x 1.8 mL. Store at 2° to 8°C
• DNA Elution Buffer 1, 1 x 15 mL. Store at RT
• DNA Elution Buffer 2, 2 x 15 mL. Store at RT
• Elution Plate, 96-well, 2 plates. Store at RT
• Wash Solution 1 Concentrate, 1 x 15 mL. Store at RT
• Wash Solution 2 Concentrate, 1 x 50 mL. Store at RT
• Multi-Sample DNA Lysis Buffer, 1 x 55 mL. Store at RT
• MicroAmp™ Clear Adhesive Film, 1 x 4 films. Store at RT
• Nuclease-Free Water, 1 x 50 mL. Store at RT
• PK Buffer, 1 x 11 mL. Store at RT
• Processing Plate, 96-well, 1 plate. Store at RT
• Proteinase K, 1 x 850 μL. Store at -15° to -25 °C
• Ribonuclease A (RNase A), 1 x 530 μL. Store at -15° to -25°C.
Format Kit
Isolation Technology Magnetic Bead
Sample Type Blood, Buccal Samples, Cells, Tissue
Final Product Type Genomic DNA
For Use With (Application) Genotyping, Real-Time Quantitative PCR (qPCR), PCR, Sequencing
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
No. of Reactions 96 Preps
Product Line MagMAX
Product Type DNA Multi-Sample Kit
Quantity 96 Preps
Target Genomic DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt