missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagMAX™ Wash Solution 1 Concentrate
Description
Invitrogen MagMAX Wash Solution 1 Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately. Supplied in one 205 mL bottle.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Buffer | Wash Buffer |
| Content And Storage | Store MagMAX™ Wash Solution 1 Concentrate at 4°C or room temperature. |
| Product Line | Ambion, MagMAX |
| Quantity | 205 mL |
| Shipping Condition | Room Temperature |
Safety and Handling
Product Identifier
- MagMAX WashSolution1Concentrate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Thiocyanic acid
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?