missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAXIscript™ SP6 Kit with Manual

Product Code. 10167004
Click to view available options
Product Type:
SP6 In Vitro Transcription Kit
SP6 In Vitro Transcription Kit with Manual
Quantity:
100 Reactions
30 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10167004

Brand: Invitrogen™ AM1310M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

SP6 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, Nuclease-free Water

Fast synthesis of RNA probes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations

Ambion MAXIscript SP6 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 30 reactions.

  • High yields of high specific activity, full-length RNA probes in just 10 minutes (RNA Yield: 2-6μg)
  • Ideal probe synthesis kit for nuclease protection assays, Northern and Southern blots, and in situ hybridization
  • Includes TURBO DNase
    Contain quality-tested DNase/RNase-free reagents
  • Synthesizes RNA probes with specific activities reaching 1 x 109 cpm/μg in just 10 minutes
  • Efficient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them perfect companion to ribonuclease protection assays
  • For synthesis of nonisotopically labeled probes
  • Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into reaction
  • For synthesis of modest amounts of unlabeled RNA (2–6μg/20μL reaction) of up to 2kb

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage SP6 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 100 Reactions
Product Line Ambion™, MAXIscript™
Product Type SP6 In Vitro Transcription Kit with Manual
Quantity 100 Reactions
Promoter SP6
Yield 2-6 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.