missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAXIscript™ SP6 Kit with Manual

Product Code. 10698265
Change view
Click to view available options
Product Type:
SP6 In Vitro Transcription Kit
SP6 In Vitro Transcription Kit with Manual
Quantity:
100 Reactions
30 Reactions
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity unitSize
10698265 SP6 In Vitro Transcription Kit with Manual 30 Reactions Each
10167004 SP6 In Vitro Transcription Kit with Manual 100 Reactions Each
10545585 SP6 In Vitro Transcription Kit 100 Reactions Each
10370255 SP6 In Vitro Transcription Kit 30 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10698265 Supplier Invitrogen™ Supplier No. AM1308M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

SP6 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, Nuclease-free Water

Fast synthesis of RNA probes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations

Ambion MAXIscript SP6 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 30 reactions.

  • High yields of high specific activity, full-length RNA probes in just 10 minutes (RNA Yield: 2-6μg)
  • Ideal probe synthesis kit for nuclease protection assays, Northern and Southern blots, and in situ hybridization
  • Includes TURBO DNase
    Contain quality-tested DNase/RNase-free reagents
  • Synthesizes RNA probes with specific activities reaching 1 x 109 cpm/μg in just 10 minutes
  • Efficient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them perfect companion to ribonuclease protection assays
  • For synthesis of nonisotopically labeled probes
  • Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into reaction
  • For synthesis of modest amounts of unlabeled RNA (2–6μg/20μL reaction) of up to 2kb

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage SP6 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 30 Reactions
Product Line Ambion, MAXIscript
Product Type SP6 In Vitro Transcription Kit with Manual
Quantity 30 Reactions
Promoter SP6
Yield 2-6 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less
What probe kits do you offer for nonisotopic probe labeling and radiolabeled probe labeling?

We offer two nonisotopic probe labeling kits:

- The BrightStar Psoralen-Biotin Nonisotopic Labeling Kit (Cat. No. AM1480)
- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)

We offer three radiolabeled probe labeling kits:

- MAXIscript Kits for in vitro transcription (Cat. Nos. AM1308 through AM1326)
- KinaseMax 5' End-Labeling Kit (Cat. No. AM1520)
- DECAprime II DNA Labeling Kit (Cat. No. AM1455)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.