missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAscript™ T3 Transcription Kit

Product Code. 10772687
Change view
Click to view available options
Quantity:
40 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10772687 40 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10772687 Supplier Invitrogen™ Supplier No. AM1338
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthesize 10 to 50 times the amount of RNA produced by conventional transcription reactions

Novel transcription reaction conditions and Ambion's patented, high-yield technology allow the synthesis of 10 to 50 times the amount of RNA produced by conventional transcription reactions. Each Ambion kit includes sufficient reagents for 40 reactions.

  • Exclusive, ultrahigh yield technology is fast: 2 hour reaction time
  • Amplifies aRNA for gene array analysis and other applications
  • Efficiently incorporates many modified nucleotides
  • A typical 20μL MEGAscript reaction with 1μg of the pTRI-Xef-1α control template will yield over 100μg of transcript
  • Versatility of the MEGAscript kit allows for manipulation of the reactions to include specialized reagents such as modified nucleotides, cap analog, or additional polymerase
Accessory Products:
MessageAmp™ aRNA Amplification Kits are powered by MEGAscript technology and have been developed especially for the amplification of cRNA prior to array analysis. To purify in vitro transcription products from free nucleotides, buffer components, and enzymes, use MEGAclear™ Kit (Mfr. No. AM1908) and the MEGAclear-96 Kit (Mfr. No. AM1909).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage T3 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 40 Reactions
Product Line Ambion, MEGAscript
Product Type T3 In Vitro Transcription kit
Quantity 40 Reactions
Promoter T3
Shipping Condition Dry Ice
Yield 100 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.