missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MICROBExpress™ Bacterial mRNA Enrichment Kit

Product Code. 10670556
Change view
Click to view available options
Quantity:
20 preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10670556 20 preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10670556 Supplier Invitrogen™ Supplier No. AM1905
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The MICROBExpress™ Bacterial mRNA Enrichment Kit is for the purification of bacterial mRNA by removing rRNA from total RNA.

The MICROBExpress™ Bacterial mRNA Enrichment Kit is for the purification of bacterial mRNA by removing rRNA from total RNA. Sufficient reagents for 20 mRNA purifications, each from 10 μg of total RNA are provided. Features of the MICROBExpress™ Kit:

  • Dramatically increase the sensitivity of array analyses and other procedures
  • Removes >95% of 16S and 23S rRNA from bacterial RNA
  • Simple procedure takes less than 2 hours
  • Works with many Gram positive and Gram negative bacteria

Rapid purification of bacterial mRNA
Isolating mRNA from bacteria has been virtually impossible, until now. The MICROBExpress™ Kit uses a novel technology to remove >95% of the 16S and 23S rRNA from total RNA of E. coli and other bacterial species. The kit is suitable for mRNA purification from a broad spectrum of Gram positive and Gram negative bacteria. The mRNA isolated with the MICROBExpress™ Kit is a superior template for synthesizing labeled cDNA for array analysis, and is ideal for quantitative RT-PCR, northern applications, and cDNA library construction. Efficient removal of 16S and 23S rRNA from bacterial RNA dramatically increases sensitivity in downstream procedures (see figure).

Start with any total RNA sample
This host-bacterial cell total RNA mixture can be obtained by a variety of RNA isolation methods. The MICROBExpress™ Kit specifically removes rRNAs; small RNAs (i.e., tRNA and 5S rRNA) are not removed. Using total RNA that lacks small RNAs as the starting material for MICROBExpress™ Kit produces the highest possible level of mRNA enrichment (see accessory products). In the first step of the MICROBExpress™ Kit procedure, total RNA is mixed with an optimized set of capture oligonucleotides that bind to the bacterial 16S and 23S rRNAs. Next, the rRNA hybrids are removed from the solution using derivatized magnetic microbeads. The mRNA remains in the supernatant and is recovered by ethanol precipitation.

Accessory products
A magnetic stand is required for use this kit. They are available in several formats, including the Single Place Magnetic Stand (Cat. No. AM10026), 6 Tube Magnetic Stand (Cat. No. AM10055), and 96-well Magnetic Stands (Cat. Nos. AM10027 and AM10050). The RiboPure™-Bacteria Kit (Cat. No. AM1925) is ideal for the purification of total RNA lacking small RNAs. Small RNAs can be removed from total RNA using the MEGAclear™ Kit (Cat. No. AM1908).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Control RNA, Capture Oligo Mix, Glycogen, and 3M Sodium Acetate at -20°C.

Store Oligo MagBeads, Binding Buffer, and Wash Solution at 4°C

Store Nuclease-free Water, 1.5mL tubes, and Collection tubes at room temperature.
Isolation Technology Hybridization Capture, Magnetic Bead
Sample Type Total RNA (Bacterial)
Final Product Type mRNA (Bacterial)
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Microarray Analysis, Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction, Northern Blotting
No. of Reactions 20 Preps
Product Line Ambion, MICROBEnrich
Quantity 20 preps
Target Total RNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.