missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Millennium™ RNA Markers-Formamide

Product Code. 10351375
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10351375 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10351375 Supplier Invitrogen™ Supplier No. AM7151

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

For accurate size determination of single-stranded RNA transcripts from 0.5 to 9kb and can be used in any Northern protocol

Sufficient markers are provided for 25 Northern gel lanes. They are a mixture of 10 discrete RNA transcripts with easy-to-remember sizes: 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9 kilobases. The markers are supplied in formamide and show increased stability and resistance to RNase compared to other large molecular weight RNA markers. They may be stained with ethidium bromide either during or after electrophoresis. Because of their quantity and even spacing, these markers are ideal for constructing very accurate standard curves for mRNA size determination. They also provide a reference scale for easy verification and comparison of known bands. 1-2μg (1-2μL) of RNA Millennium™ Markers will generate 10 distinct bands on a 1% denaturing agarose gel with ethidium bromide staining. All marker lots undergo stringent nuclease testing and are shown to be stable overnight at 37°C. Compatible with formaldehyde-containing and glyoxal-containing gels.

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at below –70°C.
Concentration 1 μg/μl
Ready to Load No
Size Range 0.5 to 9 kb
Gel Compatibility Formaldehyde-Containing Gels, Glyoxal-Containing Gels
Quantity 50 μL
Shipping Condition Dry Ice
Product Line Ambion, Millennium Markers
Product Type RNA Marker-Formamide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.