missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit

Product Code. 10596175
Change view
Click to view available options
Quantity:
50 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10596175 50 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10596175 Supplier Invitrogen™ Supplier No. AMB13455
Spotlight Offers
Use promo code 28037
View all promotions
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Generate RNA transcripts with higher protein yields

The mMESSAGE mMACHINE T7 Ultra Kit combines new cap analog, anti-reverse cap analog (ARCA), with patented high-yield transcription technology to generate RNA transcripts that produce higher protein yields compared to other transcripts upon translation.

  • RNA Yield: 20-30μg
  • Synthesize more protein from in vitro-transcribed RNA
  • Express RNA transcripts more efficiently both in vitro and in vivo
  • Stabilize transcripts in vivo with included reagents for poly(A) tailing

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage T7 Enzyme Mix, 10X Reaction Buffer 2XNTP⁄ARCA Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, and Formaldehyde Load Dye are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type ARCA Capped Synthetic mRNA, mRNA, Synthetic mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 50 Reactions
Product Line Ambion, mMESSAGE mMACHINE
Product Type T7 In Vitro Transcription kit
Quantity 50 Reactions
Promoter T7
Shipping Condition Dry Ice
Yield 20-30 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.