missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mRNA Catcher™ PLUS Purification Kit

Product Code. 10105302
Change view
Click to view available options
Quantity:
96 Preps
960 Preps
Product Type:
Buffers Only
Purification Kit
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize
10105302 960 Preps Buffers Only Each
10594823 96 Preps Purification Kit Each
10247402 960 Preps Purification Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10105302 Supplier Invitrogen™ Supplier No. K157004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

1 96-well mRNA Catcher PLUS plate; 4mL 2X Lysis Buffer L20; 30mL Wash Buffer W15; 4mL Dilution Buffer DL1; 8mL Elution Buffer E3; 100μL 0.5M DTT (4°C)

The mRNA Catcher PLUS Purification Kit enables high-throughput purification of mRNA from cells, tissues, blood, and total RNA in a 96-well format.

The mRNA Catcher PLUS Purification Kit enables high-throughput purification of mRNA from cells, tissues, blood, and total RNA in a 96-well format. By taking advantage of locked nucleic acid (LNA) technology, the mRNA Catcher PLUS kit provides improved performance and a faster protocol, while preserving mRNA yield and purity.

The mRNA Catcher PLUS Purification Kit offers:

  • Robust performance—high binding capacity per well (>80 ng/well)
  • Reproducible results—low CVs from well-to-well and plate-to-plate (∼2%)
  • Optimized protocols—for small sample sizes (100 cells, 4 mg tissue, and 40 μL blood)
  • Compatible plate design—may be used with most PCR cyclers

How it works

mRNA from total RNA, cells, tissue, and blood samples is hybridized to an immobilized, single-stranded 20-mer oligo(dT)-containing LNA. The incorporation of LNA into the oligo probe increases the hybridization efficiency and specificity of the mRNA binding. The mRNA can either be eluted or reversed-transcribed to cDNA directly in the well. The mRNA Catcher PLUS method does not require centrifugation, precipitation, phase separation, or column filtration. In addition, the entire method is amenable to automation for streamLined, high-throughput expression profiling for large-scale processes. Isolated mRNA is suitable for use in downstream applications such as RT-PCR and qRT-PCR.

Order Info

Shipping Condition: Wet ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
No. of Reactions 960 reactions
Product Type Buffers Only
Quantity 960 Preps
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.