missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NorthernMax™-Gly Kit

Product Code. 10146584
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10146584 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10146584 Supplier Invitrogen™ Supplier No. AM1946

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Positive Control RNA, pTRI-GAPDH Mouse, DECAtemplate §-actin-Mouse, Formaldehyde Load Dye, ULTRAhyb Buffer, 10X Denaturing Gel Buffer, and 10X MOPS Gel Running Buffer, Agarose-LE, Transfer Buffer, Low Stringency Wash Solution No. 1, High Stringency Wash Solution No. 2, and RnaseZap™, nuclease-free water

An alternative to formaldehyde-based systems for northern analysis and uses a glyoxal/DMSO Loading Solution for sample denaturation

  • Use of glyoxal/DMSO instead of formaldehyde for sample preparation avoids the need to pour and run gels in a fume hood, and eliminates the safety issues associated with use of formaldehyde
  • Glyoxal gels can be prepared at the lab bench: ;no noxious reagents
  • Increased sensitivity up to 100x over standard hybridization protocols
  • Reduce hybridization to 2 hours for many messages
  • Complete kit contains ULTRAhyb™ Hybridization Buffer and gel and wash reagents for 10 to 20 gels
  • Compatible with RNA, DNA, or oligonucleotide probes labeled isotopically or nonisotopically
  • Sufficient reagents are provided for processing 1000cm2 membrane
  • RNA samples denatured with glyoxal may show sharper bands on northern blots compared to samples denatured and run in the presence of formaldehyde
  • The streamlined NorthernMax-Gly procedure is more convenient and faster than the standard glyoxal gel protocol: A single reagent is used for sample denaturation and loading, the incubation time for sample denaturation has been reduced, and no recirculation of buffer is required during electrophoresis
  • The volume ratio of glyoxal denaturation solution to sample RNA is lower than many protocols, so that sample precipitation prior to gel loading is usually not required
  • An RNA sample and template for probe synthesis are also provided as controls for each kit
  • Compatible with glyoxal-containing gels

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Positive Control RNA, pTRI-GAPDH Mouse, DECAtemplate §-actin-Mouse, and Glyoxal Load Dye should be stored at -20°C.
• ULTRAhyb™ Buffer and 10X Gel Prep⁄Running Buffer should be stored at 4°C.
• Agarose-LE™, Transfer Buffer, Low Stringency Wash Solution #1, mL High Stringency Wash Solution #2, and RNaseZap™ should be stored at room temperature.
• Nuclease-free water may be stored at any temperature.
Gel Compatibility Glyoxal-Containing Gels
Quantity 1 kit
Shipping Condition Room Temperature
Product Type Northern Analysis Reagents

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.