missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phenol:Chloroform:IAA, 25:24:1, pH 6.6

Product Code. 10554645
Click to view available options
Quantity:
100 mL
400 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10554645

Brand: Invitrogen™ AM9730

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Eliminates handling problems and offers a convenient, safe, and easy alternative to preparing solutions from crystalline phenol

Ambion Acid Phenol:Chloroform:IAA (25:24:1) is premixed and supplied at pH 6.6 (pH 7.9 ± 0.2), with included buffer. Acid Phenol:Chloroform:IAA is provided in one bottle of 400 mL. In RNA and DNA extraction procedures, Acid Phenol:Chloroform:IAA (pH 7.9) helps to stabilize the interface and prevents foaming when mixing. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality tested, saturated phenols are ready-to-use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenols
Content And Storage Store at 4°C or -20°C.
Packaging Type Bottle
pH 6.6
Product Line Ambion
Purity Molecular Biology Grade
Quantity 100 mL
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.