missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ FFPE RNA Isolation Kit

Product Code. 10104492
Change view
Click to view available options
Quantity:
50 Preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10104492 50 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10104492 Supplier Invitrogen™ Supplier No. K156002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Includes

Sufficient for up to 50 reactions and each reaction is capable of purifying total RNA for up to 10 slices of 10 μm-thick FFPE sections. Kit includes: Deparaffinization/Protinase K digestion buffer, Protein/RNase Degrading buffer, 50 spin columns, 50 wash tubes, 50 recovery tubes, Binding buffer, and Wash buffer.

The PureLink FFPE RNA Isolation Kit provides a simple, reliable, and rapid method to isolate high-quality total RNA from formalin-fixed, paraffin-embedded (FFPE) tissue specimens.

The PureLink FFPE RNA Isolation Kit provides a simple, reliable, and rapid method to isolate high-quality total RNA from formalin-fixed, paraffin-embedded (FFPE) tissue specimens for retrospective basic and clinical biological studies of gene expression without the use of toxic organic solvents for deparaffinization.

Rapid and Non-Toxic Isolation Procedure
The PureLink FFPE RNA Isolation Kit employs a simple heating procedure for deparaffinization, instead of relying on the use of toxic organic solvents such as xylene. RNA isolation can be completed in 30 to 40 minutes instead of days, as is required for older FFPE isolation methods. The heating procedure for deparaffinization partly restores RNA template structure for cDNA synthesis and improves template quality for reverse transcription.

High Yield, High Quality RNA
The PureLink FFPE RNA Isolation Kit offers higher yields and purity as compared to other commercially available RNA purification systems, allowing for RT-PCR products over 1 kb in length. In addition, the PureLink FFPE RNA Isolation Kit minimizes genomic DNA contamination of the purified RNA sample and provides an option for DNase digestion.

Purified Total RNA is Suitable for Use with a Variety of Downstream Applications and Products
RNA extracted from FFPE specimens is suitable for downstream applications such as reverse transcription, real-time PCR (qPCR), real-time RT-PCR (qRT-PCR), and microarray analysis.

  • Offers a rapid and environmentally friendly procedure for the extraction of RNA from FFPE tissue specimens
  • Isolates RNA with maximum yield and minimum degradation

Not high-throughput compatible (Manual)

Applications
DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from FFPE Samples

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 15 mL Melting Buffer; store at room temperature
• 1 mL Proteinase K in buffer; room temperature
• 20 mL Binding Buffer L3; room temperature
• 20 mL Wash Buffer W5; room temperature
• 4 mL RNase-free Water; room temperature
• 50 Spin Cartridges with Collection Tubes; room temperature
• 50 Wash Tubes; room temperature
• 50 Recovery Tubes; room temperature
• 0.1 mL DNase I, Amplification Grade; -20°C
Isolation Technology Silica Spin Column
Elution Volume 50 μL
Sample Type Formalin-fixed, Paraffin-embedded (FFPE) Tissue
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 50 Preps
Scale Mini
Shipping Condition Box 1: Ice
Box 2: Room Temperature
Starting Material Amount 3–8 pieces of FFPE with 10 μm thick sections
Yield >1 μg
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.