Learn More
Invitrogen™ RiboMinus™ Human/Mouse Transcriptome Isolation Kit
Description
Includes
Contains two modules; the RiboMinus Human/Mouse Probe Module and the Concentration Module. The RiboMinus Human/Mouse Probe Module contains the RiboMinus magnetic beads, the RiboMinus human/mouse probe, and binding buffer. The Concentration Module contains spin cartridges, wash tubes, recovery tubes, binding buffer, wash buffer, and RNase-free water.
The RiboMinus™ Human/Mouse Transcriptome Isolation Kit constitutes a novel purification system that enriches the whole spectrum of mRNA species by depleting large ribosomal RNA molecules in a total RNA sample. The kit removes >95% of 18S and 28S rRNA molecules from human or mouse total RNA without relying on current polyA+ purification methods. This increases the representation of mRNA species, including non-polyA-tailed, fragmented mRNA or other regulatory RNAs that cannot be purified via polyA-selection methods. Purified RNA is suitable for several downstream applications, including, library construction, RT-PCR, and microarray.
Not high-throughput compatible (Manual)
Specifications
Specifications
| Content And Storage | The RiboMinus™ Human/Mouse Transcriptome Isolation Kit contains two modules: the RiboMinus™ Human/Mouse Probe Module and the Concentration Module The RiboMinus™ Human/Mouse Probe Module contains the RiboMinus™ magnetic beads, the RiboMinus™ human/mouse probe, and binding buffer. Store at 4°C The Concentration Module contains spin cartridges, wash tubes, recovery tubes, binding buffer, wash buffer, and RNase-free water. Store at room temperature. All reagents are guaranteed stable for 6 months when properly stored. |
| Isolation Technology | Magnetic Bead |
| Sample Type | Total RNA |
| Final Product Type | Transcriptome RNA |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| For Use With (Application) | Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting, cDNA Library Construction |
| Product Line | RiboMinus |
| Quantity | 6 preps |
| Target | Transcriptome RNA |
| System Type | RiboMinus |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.