missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ RNase H, from E. coli, 10 U/μL

Product Code. p-4918160
Click to view available options
Quantity:
1000 U
200 U
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10360395

Brand: Invitrogen™ AM2293

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Endonuclease-free, exonuclease-free, protease-free

Ribonuclease H (RNase H) is isolated from an E. coli strain that over-expresses the gene. RNase H specifically degrades RNA in RNA:DNA hybrids to produce 3'-hydroxyl and 5'-phosphate terminated products.

  • Will not degrade DNA or unhybridized RNA
  • Integral part of most RNA amplification and NASBA protocols and can be used to degrade specific RNAs when complementary DNA oligo is hybridized, such as poly(A) tail removal from mRNA hybridized to oligo(dT)
  • Rigorously tested for contaminating nonspecific endonuclease, exonuclease, RNase, and protease activity
  • Supplied in one tube containing 200U (10U/μL
  • Unit Definition: One unit of Ribonuclease H is amount of enzyme required to increase fluorescence 1.5 RFUs per sec. at 37°C using 20pmol of RNaseAlert™ probe coupled to 1000pmol of a complementary oligonucleotide as substrate

PCR & Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Content And Storage -20°C
Shipping Condition Dry Ice
Enzyme RNase
Quantity 1000 U
Product Type RNase H
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.