missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RiboPure™ RNA Purification Kit

Product Code. 10107824
Change view
Click to view available options
Product Type:
Kit
Kit with manual
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10107824 Kit Each
10321325 Kit with manual Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107824 Supplier Invitrogen™ Supplier No. AM1924
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Ambion™ RiboPure™ Kit is designed for rapid purification of high-quality RNA from tissue samples or cultured cells. The kit combines TRI Reagent™ with glass fiber filter purification to yield exceptionally pure RNA, free of residual proteins and lipids.

The Ambion™ RiboPure™ Kit is designed for rapid purification of high-quality RNA from tissue samples or cultured cells. The kit combines TRI Reagent™ with glass fiber filter purification to yield exceptionally pure RNA, free of residual proteins and lipids. The kit includes sufficient reagents for 50 purifications from 1–100 mg of tissue or 0.1–20 x 106 cultured cells each.

  • Yields the highest purity RNA attainable
  • Ideal for the most stringent downstream applications
  • Handles problematic samples with ease
  • Fast (less than 30 minutes) and easy procedure

RNA isolated using the RiboPure™ Kit is suitable for use in all common downstream applications, including cDNA synthesis, real-time and end-point RT-PCR, microarray analysis, northern blots, and RNase protection assays. The RiboPure™ protocol is especially suited for processing samples that are difficult to disrupt, or samples that have large amounts of ribonuclease or other contaminants. Samples are first homogenized in TRI Reagent™, a monophasic solution containing phenol and guanidine thiocyanate, to rapidly lyse cells and inactivate nucleases. Addition of bromochloropropane (BCP) results in separation of the homogenate into aqueous and organic phases. RNA partitions to the aqueous phase, while DNA and protein remain in the interphase and organic phase. The aqueous phase is removed and passed through a glass fiber filter that binds the RNA. The filter is washed, and the RNA is eluted. The procedure is compatible with tissues that have been stored in Ambion's RNAlater™. Total RNA yield is typically 100–500 μg per 100 mg of tissue, depending on the type of tissue.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 50 mL TRI Reagent; store at 4°C
• 50 Filter Cartridges; room temperature
• 100 Collection Tubes (2 mL); room temperature
• 60 mL Wash Solution (concentrated); room temperature
• 11 mL Elution Buffer; room temperature
Isolation Technology Spin Column (Glass Fiber Filter)
Purification Time 30 min. (After sample homogenization)
Elution Volume 100 μL
Sample Type Cells, Tissue
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Product Type Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Starting Material Amount Cells: 0.1–20 x 106
Tissue: 5–100 mg
Yield Up to 500 μg
Show More Show Less
I'm getting poor quality/poor purity of RNA after isolation with the RiboPure RNA Purification kit. What could be the cause of this?

Poor quality/poor purity RNA is typically due to the following:

- Partially degraded RNA: Use fresh tissue or cells, or use RNAlater Stabilization Solution if tissue cannot be frozen immediately.
- DNA contamination: If the sample contains organic solvents, strong buffers, or an alkaline solution, we recommend performing phase separation at 8 degrees C.
- A low A260/A280 ratio (<1.6): This indicates that an insufficient amount of the TRI Reagent was used. Incubation of the homogenate at room temperature for 5 minutes can help nucleoproteins dissociate from RNA. Phenol contamination could also cause a low absorbance ratio.

What is the column cut-off size for the RiboPure RNA Purification Kits?

The column cut-off size is 200 nt. Using a RiboPure kit is not appropriate for miRNA recovery. For miRNA isolation, we would suggest using a mirVana kit, the TRIzol Plus RNA Purification Kit, or the PureLink RNA Micro Kit.

What are the differences in technology between the PureLink RNA Mini Kit and RiboPure RNA Purification Kits?

The PureLink RNA Mini Kit provides rapid column-based purification of total RNA, without organic lysis (phenol/chloroform). You can obtain up to 1,000 µg of purified RNA from a single extraction. The RiboPure RNA Purification Kits combine a phenol/guanidine thiocyanate solution with a glass-fiber filter purification method.

What is the composition of the elution buffer included in the RiboPure RNA Purification Kit (Cat. No. AM1924)?

The elution buffer composition is proprietary.

compliance-icons compliance-icons compliance-icons
Product Identifier
  • TRI Reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Phenol, Thiocyanic acid, compound with guanidine

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.