missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNA 6000 Ladder

Product Code. 10107684
Change view
Click to view available options
Quantity:
3 tubes
Unit Size:
60µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10107684 3 tubes 60µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10107684 Supplier Invitrogen™ Supplier No. AM7152
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

A set of six RNA transcripts with lengths of 0.2, 0.5, 1.0, 2.0, 4.0, and 6.0kb

The 0.2Kb band is at a concentration of 20ng/μL (A260). The RNA 6000 Ladder is designed for use with Agilent's lab-on-a-chip. Three tubes containing 20μL each are provided. 150ng (1μL) will generate 6 distinct bands when run on a RNA Labchip™ using the Total RNA or mRNA assay. Compatible with RNA LabChip.

DNA and RNA Purification and Analysis, Nucleic Acid Quantitation, RNA Quantitation

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at below –70°C.
Concentration 150 μg/ml
Ready to Load No
Size Range 0.2 to 6 kb
Gel Compatibility RNA LabChip
Quantity 3 tubes
Shipping Condition Dry Ice
Product Line Ambion
Product Type RNA 6000 Ladder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.