Learn More
Invitrogen™ RNAqueous™-96 Automated Kit
Description
Includes
4 Filter Plates, 4 Collection Plate with lids, 4 Culture Plates, 1 Heat Plate with lid, 100mL Nuclease-free Water, 100mL Elution Solution, 100mL Lysis/Binding Solution, 100mL Rebinding Mix, and 2 x 175mL Wash Solution Concentrate, (1.25mL DNase I (20U/μL) and 12mL DNase 1 Buffer
The Invitrogen RNAqueous-96 Automated Kit is used for automated, phenol-free total RNA isolation from 100-500,000 cells or 0.1–1 mg tissue using a guanidinium-based lysis/denaturant and glass fiber filter separation technology. The kit contains enough reagents for 384 (4 x 96) RNA purifications. The RNAqueous-96 Automated Kit uses an RNA-binding glass-fiber filter to provide high, reproducible yields of intact RNA. An optional on-the-filter DNase treatment can be performed to ensure removal of genomic DNA for RT-PCR applications.
Spezifikation
Spezifikation
| Content And Storage | • 4 Filter Plates; store at room temperature • 4 Collection Plates with lids; room temperature • 4 Culture Plates; room temperature • 1 Heat Plate with Lid; room temperature • 100 mL Nuclease-free Water; room temperature • 100 mL Elution Solution; room temperature • 100 mL Lysis/Binding Solution; 4°C • 100 mL Rebinding Mix; 4°C • 2 x 175 mL Wash Solution (concentrated); 4°C • 1.25 mL DNase 1; -20°C • 12 mL DNase 1 Buffer; -20°C |
| Isolation Technology | Vacuum Filter Plate (Glass Fiber Filter) |
| Elution Volume | 105 μL |
| Sample Type | Cells, Tissue |
| Final Product Type | Total RNA |
| High-throughput Compatibility | High-throughput Compatible |
| For Use With (Equipment) | MultiPROBE™ II HT, Biomek™ 2000, Liquid Handling Systems |
| For Use With (Application) | RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling |
| Quantity | 384 Preps |
| Shipping Condition | Box 1: Dry Ice Box 2: Room Temperature |
| Mehr anzeigen |
For Research Use Only. Not for use in diagnostic procedures.
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.