missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ RNase A, affinity purified, 1 mg/mL

Product Code. 10095064
Change view
Click to view available options
Quantity:
1 mg
200 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10095064 200 μg Each
10544255 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10095064 Supplier Invitrogen™ Supplier No. AM2270
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity

Ambion RNase A is endonuclease that specifically cleaves 3' of U and C residues. Affinity Purified RNase A is intended for critical applications when absence of DNase and other nonspecific nuclease activities is essential.

  • Supplied in one tube containing 200μg (1mg/mL)
  • Preparation contains RNase B, a carbohydrate isoform of RNase A

DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Content And Storage -20°C
Shipping Condition Dry Ice
Enzyme RNase
Quantity 200 μg
Product Type RNase A
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.