missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ RNase I, cloned, 100 U/μL

Product Code. 10545195
Change view
Click to view available options
Quantity:
10,000 U
25,000 U
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10545195 10,000 U Each
10330065 25,000 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10545195 Supplier Invitrogen™ Supplier No. AM2294
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Efficiently cleaves after all four bases of single-stranded RNA, in contrast to RNase A, which only cleaves after C and U residues

RNase I degrades all RNA dinucleotide bonds leaving a 5' hydroxyl and 2', 3' cyclic monophosphate. RNase I will degrade any RNA to mixture of mononucleotides, dinucleotides, and trinucleotides and does not degrade DNA, although it will bind to DNA. It has marked preference for single-stranded RNA over double-stranded RNA, which allows it to work well in ribonuclease protection assays.

  • Supplied in one tube of 10,000U (100U/μL)
  • Endonuclease-free, exonuclease-free, protease-free
  • Rigorously tested for nonspecific contaminating endonuclease, exonuclease, and protease activity
  • Unit Definition: One unit is amount of enzyme required to produce 1μg of acid-soluble material from mouse liver RNA in 30 min. at 37°C

DNA & RNA Purification & Analysis, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 100 U/μL
Content And Storage -20°C
Shipping Condition Dry Ice
Enzyme RNase
Quantity 10,000 U
Product Type RNase I
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.