missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SDS, 10% Solution, RNase-free

Product Code. 10525975
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10525975 500 mL Each
10712687 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10525975 Supplier Invitrogen™ Supplier No. AM9822

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Molecular biology grade, Invitrogen SDS, 10% Solution is supplied in one bottle containing 500 mL.

Molecular biology grade, Invitrogen SDS, 10% Solution is supplied in one bottle containing 500 mL. The buffer is certified RNase-free, economical, and ready-to-use. SDS is used as a component of both hybridization and wash buffers for northern and Southern blots and has many other uses around the lab.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Concentration 0.1
Packaging Type Bottle
Product Type SDS
Product Line Ambion
Quantity 500 mL
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.