missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSPE (20X), RNase-free

Product Code. 10187344
Change view
Click to view available options
Quantity:
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10187344 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10187344 Supplier Invitrogen™ Supplier No. AM9767
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Molecular biology grade, Invitrogen 20X SSPE solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use.

Molecular biology grade, Invitrogen 20X SSPE solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Concentration 20 X
Volume (Metric) 1 L
Form Liquid
For Use With (Application) Nucleic Acid Gel Electrophoresis and Blotting
Purity or Quality Grade Molecular Biology Grade
Quantity 1 L
Shipping Condition Room Temperature
Product Line Ambion
Product Type Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.