missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SUPERase·In™ RNase Inhibitor (20 U/μL)

Product Code. 10085994
Change view
Click to view available options
Quantity:
10,000 U
2500 U
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10085994 2500 U Each
10773267 10,000 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10085994 Supplier Invitrogen™ Supplier No. AM2694
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Supplied in a single tube

Binds and inactivates a wide variety of RNases

SUPERase·In™ RNase Inhibitor is a protein-based inhibitor of nonhuman origin that noncovalently binds and inhibits the most common and troublesome RNases, including RNase A, B, C, 1, and T1. SUPERase·In RNase Inhibitor can be used in any application where RNase contamination could be problematic. Because it inhibits a broader range of RNases than traditional RNase inhibitors, SUPERase·In is the most effective RNase inhibitor available, providing a higher level of protection against degradation. SUPERase·In does not interfere with other enzymes such as RNA polymerases, reverse transcriptase, or Taq DNA polymerase. Additionally, SUPERase In is active up to 65°C and over a pH range of 5.5 to 8.5.

  • Completely removes RNase contamination from glass and plastic surfaces
  • Excels at removing high levels of RNase contamination where similar products fail
  • Proven effective at removing high concentrations of dried-on RNase A
  • Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free
Unit definition
SUPERase·In RNase Inhibitor at 1U/μL will block the degradation of 0.1μg/μL labeled RNA by 2.5pg/μL of RNase A, 2.5pg/μL of RNase I, and 0.0075U/μL of RNase T1, for four hours at 37°C, in 20mM Tris-HCl, pH 7.5, 50mM NaCl, 1mM EDTA. Analysis is by denaturing PAGE. SUPERase·In is currently the only ribonuclease inhibitor for which the unit activity is defined by such a functional assay.

If using this product with standard antibody purification methods, please contact technical support to discuss experimental design.

Gene Expression Analysis and Genotyping, In Vitro Transcription, PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 50 mM
Content And Storage Supplied in a single tube that should be stored at -20°C.

Storage buffer: 2 mM KH2PO4, 8 mM Na2HPO4,2.7 mM KCl, 137 mM NaCl, pH 7.4 in 50% glycerol
Product Line Ambion, SUPERase•In
Quantity 2500 U
Shipping Condition Dry Ice
What temperature and pH is SUPERase·In RNase Inhibitor (Cat. No. AM2694, AM2696) active up to?

SUPERase·In RNase Inhibitor (20 U/µL) (Cat. No. AM2694, AM2696) is active up to 65 degrees C and over a pH range of 5.5 to 8.5.

What kind of Rnases can be inhibited using SUPERase.In RNase Inhibitor?

SUPERase.In RNase Inhibitor is a protein-based inhibitor of nonhuman origin that noncovalently binds and inhibits the most common and troublesome RNases, including RNase A, B, C, 1, and T1.

What storage buffer do you recommend for the SUPERase.In RNase Inhibitor?

The storage buffer we recommend is as follows: 2 mM KH2PO4, 8 mM Na2HPO4, 2.7 mM KCl, 137 mM NaCl, pH 7.4 in 50% glycerol.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.