missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ T4 RNA Ligase, cloned, 5 U/μL

Product Code. 10126774
Change view
Click to view available options
Quantity:
2,500 units, 5 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10126774 2,500 units, 5 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10126774 Supplier Invitrogen™ Supplier No. AM2141
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Catalyzes formation of a phosphodiester linkage between a 5'-phosphoryl-terminated nucleic acid donor and a 3'-hydroxyl-terminated nucleic acid acceptor

  • Substrates include RNA, DNA, and oligonucleotides. One tube containing 500U (5U/μL) is provided along with 10X Reaction Buffer
  • The enzyme can be used for tagging the 5′ ends of mRNA with oligonucleotides for mapping studies (RACE), 3′-end labeling RNA molecules, and circularizing RNA and DNA molecules
  • Unit Definition: One unit catalyzes the formation of 1nmol of [5′-32P]-rA12-18 into a phosphatase-resistant form in 30 minutes at 37°C

DNA & RNA Purification & Analysis, Nucleic Acid Labeling & Oligo Synthesis, Oligo Labeling, RNA Labeling

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage T4 RNA Ligase and 10X Reaction Buffer should be stored at –20°C.
Format Tube
Enzyme Ligase
Fidelity (vs. Taq) 10 X
No. of Reactions 250
Polymerase SP6 RNA Polymerase
Product Line Ambion
Product Type T4 Rna Ligase
Quantity 2,500 units, 5 μL
Shipping Condition Dry Ice
Source RNA
Form Solution
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.