missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T7-Oligo(dT)24V Promoter Primer, Anchored (100 ng/μl)

Product Code. 10311045
Change view
Click to view available options
Quantity:
40 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10311045 40 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10311045 Supplier Invitrogen™ Supplier No. AM5712
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

The primer is supplied in a single tube.

For use in reverse transcription reactions to append a T7 RNA polymerase promoter to cDNA fragments

Each Invitrogen primer is PAGE purified and performance tested in reverse transcription and subsequent in vitro transcription reactions. Promoter primers are anchored with a G, A, or C (V).

Gene Expression Analysis and Genotyping, In Vitro Transcription, PCR and Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Primer
Content And Storage The primer is supplied in a single tube and should be stored at –20°C.
Shipping Condition Dry Ice
Product Line Ambion
Quantity 40 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.