missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TaqMan™ Cells-to-CT™ Control Kit

Product Code. 10150934
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10150934 100 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10150934 Supplier Invitrogen™ Supplier No. 4386995
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Xeno RNA Control, 20X Xeno RNA Control TaqMan Gene Expression Assay, and 20X ACTB (β-actin) TaqMan Gene Expression Assay for 100 reactions.

The Invitrogen™ TaqMan™ Cells-to-CT Control Kit is designed for use with the TaqMan™ Gene Expression Cells-to-CT Kit.

The Invitrogen™ TaqMan™ Cells-to-CT Control Kit is designed for use with the TaqMan™ Gene Expression Cells-to-CT Kit. The Control Kit includes XenoRNA™ Control, a synthetic RNA transcript with a unique sequence that lacks homology to current annotated biological sequences, making it an ideal control for any experiment. The Control Kit also includes a TaqMan™ Gene Expression Assay for the XenoRNA™ Control target, and a TaqMan™ Gene Expression Assay for the highly expressed endogenous control gene β-actin.

  • Confirms sufficient cell input
  • Monitors the presence of RT or real-time PCR inhibitors
  • Controls for sample normalization
  • Monitor inhibition and confirm sufficient cell input
  • Evaluate compatibility with new cell lines
  • Internal Probe Modification: Non-Fluorescent Quencher (3')FAM (5')MGB (Minor Groove Binder) (3')

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Xeno™ RNA Control, 20X Xeno™ RNA Control Taqman™ Gene Expression Assay, and 20X ACTB (β-actin) TaqMan™ Gene Expression Assay are all stored at -20°C.

All reagents are guaranteed stable for 6 months when properly stored.
Control Template XenoRNA™ control (synthetic RNA transcript)
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Gene Symbol ACTB
Internal Probe Modification MGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Label or Dye FAM
Product Line Ambion, Cells-to-CT, TaqMan
Product Type Cells-to-C Control Kit
Quantity 100 reactions
Shipping Condition Dry Ice
Sufficient For 100 Reactions
Target XenoRNA™ control, β-actin
For Use With (Application) Gene Expression
No. of Reactions 100 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.