missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TaqMan™ Fast Cells-to-CT™ Kit

Product Code. 10712697
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10712697 100 reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10712697 Supplier Invitrogen™ Supplier No. 4399003
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Stop Solution: 500μL tube; DNase I: 55μL tube; 20X RT Enzyme Mix: 275μL tube; Lysis Solution: 5.5mL bottle; 2X RT Buffer: 5.5mL bottle; TaqMan Fast Universal PCR Master Mix: 4 x 1.25mL bottle

The TaqMan™ Fast Cells-to-CT™ Kit makes it possible to use Fast-cycling, real-time PCR chemistry and instrumentation to perform gene expression analysis directly from cultured cells without RNA purification.

  • Complete: Optimized workflow includes cell lysis reagents with gDNA removal, RT enzyme mix, RT buffer, and TaqMan™ Fast Universal PCR Master Mix
  • Fast: Prepare samples at room temperature in 7 minutes, including DNase treatment; TaqMan™ Fast Universal PCR Master Mix delivers real-time PCR results in ∼35 minutes
  • Convenient: Lyse samples in tube or directly in 96- or 384-well culture plates
  • Robust: Perform gene expression analysis on 10 to 100,000 cells per sample, with results equivalent to those from purified RNA
  • Efficient: Contains sufficient reagents to generate 500 real-time PCR results from 100 starting samples

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Contains:
• Stop Solution: 500 μL tube
• DNase I: 55 μL tube
• 20X RT Enzyme Mix: 275 μL tube
• Lysis Solution: 5.5 mL bottle
• 2X RT Buffer: 5.5 mL bottle
• TaqMan™ Fast Universal PCR Master Mix: 4 x 1.25 mL bottle

Store Stop Solution, DNase I, and 20X RT Enzyme Mix at -20°C.
Store Lysis Solution, 2X RT Buffer, and TaqMan™ Fast Universal PCR Master Mix at 4°C.

Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance Low
PCR Method 2-step RT-qPCR
Polymerase DNA Polymerase
Reaction Speed Fast
For Use With (Equipment) 7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Passive Reference Dye ROX (Pre-mixed)
Product Line Ambion, Cells-to-CT, TaqMan
Product Type Fast Cells-to-C Kit
Quantity 100 reactions
Reverse Transcriptase M-MLV
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) Gene Expression
No. of Reactions 100 reactions
Sample Type Cell
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.