missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TaqMan™ Gene Expression Cells-to-CT™ Kit

Product Code. 10664844
Change view
Click to view available options
No. of Reactions:
100 Reactions
40 Reactions
400 Reactions
Unit Size:
100 reactions
400 reactions
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10664844 400 Reactions 400 reactions
10147254 40 Reactions Each
10625614 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 10664844 Supplier Invitrogen™ Supplier No. AM1729

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

The TaqMan™ Gene Expression Cells-to-CT™ Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification.

The TaqMan™ Gene Expression Cells-to-CT™ Kit makes it possible to perform expression analysis directly from cultured cells without RNA purification. This kit saves time and offers a simple workflow that is suitable for a few samples or can be easily incorporated into automated, high-throughput applications.

The TaqMan™ Gene Expression Cells-to-CT Kit is:
Complete—optimized workflow includes cell lysis reagents with gDNA removal, RT enzyme mix, buffer, and new TaqMan™ Gene Expression Master Mix
Fast—7-minute sample prep, including DNase treatment, at room temperature
Easy—lyse samples in a tube or directly in culture plates
Robust—perform gene expression analysis on 10–100,000 cells per sample; results equivalent to those from purified RNA
Efficient—contains sufficient reagents to generate 500 real-time PCR results from 100 starting samples

Featuring a unique method for lysing cultured cells while removing genomic DNA and preserving RNA integrity, the TaqMan™ Gene Expression Cells-to-CT™ Kit contains reverse transcription (RT) reagents for cDNA synthesis and TaqMan™ Gene Expression Master Mix for real-time PCR analysis. TaqMan™ primer⁄probe sets are sold separately.

Simple 7-minute sample preparation: part of a complete workflow
Whether you are using plates or tubes, the TaqMan™ Gene Expression Cells-to-CT™ Kit, which uses the simple 7-minute sample preparation procedure outlined (see figure), is designed for 10–100,000 cultured cells⁄sample. Cells are washed in PBS and lysed in solution for 5 minutes at room temperature; DNase treatment can be performed simultaneously. Lysis is terminated at room temperature by a 2-minute incubation with Stop Solution. The lysates are now ready for reverse transcription or storage at –20°C. Because samples can be processed directly in culture wells (96 or 384 wells), sample handling and the potential for sample loss or transfer error are minimized, facilitating rapid, high-throughput processing. Unlike old-fashioned, multi-step RNA isolation protocols, no heating, washing, or centrifugation steps are required. The kit greatly simplifies a laborious 30–60 minute process and reduces it to 7 minutes.

The TaqMan™ Gene Expression Cells-to-CT™ Kit workflow enables unsurpassed gene expression evaluation with any of the >700,000 TaqMan™ Gene Expression Assays. This kit has been extensively tested for specificity with a broad selection of TaqMan™ Gene Expression Assays and shows performance equivalent to that obtained with purified RNA (see figure).

Achieve unsurpassed performance and sensitivity
Unlike some competitor kits that limit the amount of lysate in the RT reaction to 5%, the TaqMan™ Gene Expression Cells-to-CT™ Kit can accommodate 45% of the total RT reaction volume as cell lysate. Additionally, cDNA can comprise up to 45% of the real-time PCR reaction volume. The large lysate volume in the optimized RT reaction, along with the large cDNA volume in the subsequent real-time PCR using the TaqMan™ Gene Expression Master Mix, lead to maximum sensitivity. The master mix amplifies the target precisely and accurately, enabling the detection of small quantities of target, such as transcripts expressed at low levels.

The ability to detect limited target quantities was tested by mixing a constant number of human cells with various numbers of mouse cells. The cell mixtures were prepared using the TaqMan™ Gene Expression Cells-to-CT™ and assayed for a mouse-specific and human-specific gene. Comparative data were generated in the same manner with RNA purified by traditional methodology. The data show that RNA from as few as 10 mouse cells was detectable in a background of RNA from 10,000 human cells (see figure). The ability of the TaqMan™ Gene Expression Cells-to-CT™ Kit to detect relative low abundance transcripts was equivalent to that of purified RNA; at low levels, it was superior.

Additionally, the performance of the TaqMan™ Gene Expression Cells-to-CT™ Kit was compared to competitor lysate kits and to purified RNA. Inputs of 100–100,000 cells⁄lysis reaction were examined. The sensitivity of the TaqMan™ Cells-to-CT™ Kit protocol was equivalent to that obtained with purified RNA, and it surpassed competitor lysates (see figure). Furthermore, the lack of inhibition at high cellular inputs and the low variation among technical replicates demonstrate the reliability of this approach for gene expression studies using cultured cells.

Proven performance, proven together
All components of the TaqMan™ Gene Expression Cells-to-CT™ Kit have been optimized for consistent and reliable performance. This removes the guesswork involved in assembling separate sample preparation, RT, and real-time PCR kits. And the TaqMan™ Gene Expression Cells-to-CT™ Kit has been validated with TaqMan™ Gene Expression Assays for added quality assurance.

Accessory product
The TaqMan™ Cells-to-CT™ Control Kit was designed for use with the TaqMan™ Gene Expression Cells-to-CT™ Kit. This kit contains an endogenous control (ACTB) to normalize for sample loading variability and an artifical XenoRNA™ control and corresponding TaqMan™ Gene Expression Assay to monitor the effects of PCR inhibition.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
• Stop Solution: 2 × 1 mL tubes
• DNase I: 220 μL tube
• 20X RT Enzyme Mix: 1.1 mL tube
• Lysis Solution: 22 mL bottle
• 2X RT Buffer: 22 mL bottle
• TaqMan™ Gene Expression Master Mix: 4 × 5 mL bottles

Store Stop Solution, DNase I, and 20X RT Enzyme Mix at -20°C.
Store Lysis Solution, 2X RT Buffer, and TaqMan™ Gene Expression Master Mix at 4°C.
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR
Polymerase DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7900HT System, Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Passive Reference Dye ROX (Pre-mixed)
Product Line Ambion, Cells-to-CT, TaqMan
Product Type Gene Expression Cells-to-C Kit
Purity or Quality Grade UP (Ultra Pure)
Quantity 400 reactions
Reverse Transcriptase M-MLV
Shipping Condition Dry Ice
Sufficient For 400 Reactions
No. of Reactions 400 Reactions
Reagent Type Cell Lysis
Sample Type Cell
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.