missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TaqMan™ MicroRNA Cells-to-CT™ Kit

Product Code. 10370845
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10370845 100 reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10370845 Supplier Invitrogen™ Supplier No. 4391848
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Includes cell lysis reagents, DNase, TaqMan MicroRNA RT reagents, and TaqMan Universal PCR Master Mix. 100 sample prep rxns, 100 DNase rxns, 100 reverse transcription rxns, and 500 20μL PCR rxns for 100 reactions.

First to offer a simple, complete workflow for miRNA expression analysis from a few samples, or it can easily be incorporated into automated, high throughput applications.

First to offer a simple, complete workflow for miRNA expression analysis from a few samples, or it can easily be incorporated into automated, high throughput applications.

  • Complete, validated solution: Optimized workflow includes cell lysis reagents, DNase, TAqMan MicroRNA RT reagents, and TAqMan Universal PCR Master Mix
  • Fast, simple, and convenient: Prepare samples in 10 minutes; eliminate tedious RNA isolation
  • Gold standard specificity: Achieve superior results with TAqMan MicroRNA reagents
  • Robust performance: Results equivalent to purified RNA
  • Broad input dynamic range: Linear detection from 10 to 100,000 cells
  • Fewer resources used, less hazardous, less waste

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Includes cell lysis reagents, DNase, TaqMan™ MicroRNA RT reagents, and TaqMan™ Universal PCR Master Mix.

• 100 sample prep reactions
• 100 DNase reactions
• 100 reverse transcription reactions,
• 500 x 20 μL PCR reactions

Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method 2-step RT-qPCR
Polymerase DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) 7000 System, 7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system
Passive Reference Dye ROX (Pre-mixed)
Product Line Ambion, Cells-to-CT, TaqMan
Product Type MicroRNA Cells-to-C Kit
Quantity 100 reactions
Reverse Transcriptase M-MLV
Shipping Condition Dry Ice
Sufficient For 100 Reactions
For Use With (Application) miRNA analysis
No. of Reactions 100 Reactions
Sample Type Cell
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.