missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THE RNA Storage Solution

Product Code. 10449224
Change view
Click to view available options
Quantity:
50 mL
10 x 1 mL
Unit Size:
10mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10449224 10 x 1 mL 10mL
10659075 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10449224 Supplier Invitrogen™ Supplier No. AM7000

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The Invitrogen RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer.

The Invitrogen RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer. This 1 mM sodium citrate, pH 6.4 ± 0.2 buffer is provided in one bottle containing 50 mL.

The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. The RNA Storage Solution has two features that minimize base hydrolysis of RNA: a low pH, and sodium citrate, which is an efficient chelating agent. The RNA Storage Solution is compatible with all common RNA applications such as reverse transcription, in vitro transcription, northern analysis, and nuclease protection assays. The RNA Storage Solution is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mM
Content And Storage Store at -20°C.
Shipped at ambient temperature.
Packaging Type Tube
Product Line Invitrogen
Product Type RNA Storage Reagent
Quantity 10 x 1 mL
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.