missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thin-walled, frosted lid, RNase-free PCR tubes (0.5 mL)

Product Code. 10489034
Change view
Click to view available options
Quantity:
1000 Tubes
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10489034 1000 Tubes Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10489034 Supplier Invitrogen™ Supplier No. AM12275
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Thin-walled, flat, frosted cap provides a suitable surface for detailed labeling. 0.5 mL tubes.

Thin-walled, flat, frosted cap provides a suitable surface for detailed labeling. 0.5 mL tubes. Supplied in box of 1000. Each lot of tubes is certified RNAse- and DNase-free.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Volume (Metric) 0.5 mL
Quantity 1000 Tubes
For Use With (Equipment) Thermal Cyclers
Product Line Ambion
Material Polypropylene
Content And Storage Store at room temperature.
Purity or Quality Grade RNase-Free, DNase-Free
Reaction Speed Fast
Shipping Condition Room Temperature
Closure Type Flat Cap
Closure Included Yes
Format Tube
Product Type RNase-free PCR Tube
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.