missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ULTRAhyb™ Ultrasensitive Hybridization Buffer

Product Code. 10289154
Change view
Click to view available options
Quantity:
125 mL
4 x 125 mL
Unit Size:
500mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10289154 125 mL Each
10792117 4 x 125 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10289154 Supplier Invitrogen™ Supplier No. AM8670
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Proprietary blend of hybridization accelerators and blocking agents that greatly enhance the levels of hybridization

  • Signals that once took days to visualize are now apparent in hours
  • It is supplied in four bottles containing 125mL each
  • Increase the sensitivity of blot hybridization experiments
  • Increase random-primed DNA probe sensitivity up to 100x; RNA probe sensitivity 20x
  • Decrease hybridization time to 2 hours
  • Using standard hybridization buffers, only 1 to 5% of target molecules on a blot hybridizes to probe, making blots a relatively insensitive method for nucleic acid analysis (Vernier et al. (1996) Anal Biochem 235: 11-19); With ULTRAhyb Ultrasensitive Hybridization Buffer, the hybridization reaction approaches completion, so that as few as 10,000 molecules can be detected
  • Compatible with RNA and DNA probes labeled isotopically and nonisotopically in northerns, Southerns, and dot/slot blots
  • Contains 50% formamide and is compatible with positively charged membranes
  • Requires 10-100x less nonisotopic DNA probe than standard hybridization buffers, making it possible to perform many more hybridizations with a single labeling reaction

Applications:

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Buffer Hybridization Buffer
Quantity 125 mL
Shipping Condition Room Temperature
Product Type Ultrasensitive Hybridization Buffer
With the ULTRAhyb Ultrasensitive Hybridization Buffer, why am I getting cross-hybridization?

The extreme sensitivity of ULTRAhyb Ultrasensitive Hybridization Buffer may detect RNAs that are not the expected full-length target. Although much of the probe binding can be legitimate (hybridization to alternatively spliced, partially degraded, or closely related mRNAs), some might be hybridization to RNAs with only partial homology to the target. We recommend using a more specific probe if possible. Here are other possible causes for cross-hybridization and solutions offered:
- The hybridization stringency may have been inadequate. Increasing the hybridization and wash temperature 3-10 degrees C can greatly reduce the levels of non-target hybridization. Simply reducing the amount of time used to expose the blot to film might also alleviate the problem.

- The probes might have contained non-target sequence. The presence of vector sequence within the probe can cause hybridization to RNAs sharing sequence homology with the vector. If the probe template contains vector sequence, cleave it by restriction digestion and then gel purify the sequence of interest before labeling.

- Too much non-isotopic probe may have been present. Non-isotopic probes can have problems with cross-hybridization, especially when they are used at 42 degrees C. We have observed that lowering the probe concentration 10 to 100-fold in the hybridization reaction will greatly reduce non-specific hybridization while having little if any impact on target-specific hybridization.

With the ULTRAhyb Ultrasensitive Hybridization Buffer, why am I getting low signal on my blot?

Here are possible causes and solutions:
- Not enough probe (or label) was used. Using less than the recommended amounts of probe, using low specific activity probe, or using less than full-length probes can lead to low signal. We recommend checking each of these factors if low sensitivity is observed.

- The hybridization and washes may have been too stringent. We recommend lowering the hybridization temperature or the wash time and temperature. This may be especially helpful for oligonucleotide probes. Note that reducing stringency can lead to higher background and cross-hybridization.

With the ULTRAhyb Ultrasensitive Hybridization Buffer, why am I getting high background on my blot?

Here are possible causes and solutions:
- There could be precipitates in the ULTRAhyb Ultrasensitive Hybridization Buffer. Inadequate solubilization of the hybridization buffer is one of the primary causes of high background. We recommend increasing the amount of time used to preheat the buffer and ensure that there is no precipitate in the buffer before adding it to the blot. ULTRAhyb Ultrasensitive Hybridization Buffer may start to precipitate at temperatures below 25-30 degrees C.

- Prehybridization may have been inadequate. We recommend increasing the blot prehybridization time from 30 min to 1 hr.

- The probe may have been too old. We recommend using fresh probe. Isotopic probes that are several days old tend to produce higher background than freshly prepared probes. This is attributed to probe size; radiolytic decay reduces the size of the probe molecules over time.

- Unincorporated radionucleotides may have been present. Although we don't ordinarily recommend removing free nucleotides, we have occasionally observed high background from unincorporated label. We recommend removing free label by precipitation (0.5 M NH4OAc and 2 volumes EtOH) or with a spin column designed for this purpose.

- Hybridization stringency may have been inadequate. We recommend the following:
-If hybridizing at 42 degrees C, try raising the hybridization temperature to 4 degrees C, or even to 55 degrees C.
-If hybridizing at 68 degrees C, hybridization stringency is unlikely to be causing background. In our experience, raising the hybridization temperature above 68 degrees C does not decrease background.

- Ionic interactions could have led to high background. If the background signal makes the blot look uniformly dark, we recommned adding a high salt wash to minimize ionic interactions between the probe and the hybridization membrane. To do this, after the ordinary washes, add a 2 x 15 min wash in 5X SSC or SSPE, 0.5% SDS at 68 degrees C for RNA probes, or at 60 degrees C for DNA probes.

- Washing may have been inadequate. We recommend the following:
-Double check that your wash buffers contain SDS. Wash buffers lacking SDS are not recommended for use with ULTRAhyb Ultrasensitive Hybridization Buffer. Doubling the wash times and/or washing at higher temperatures can reduce background. Wash temperatures can be raised from 42 degrees C to 55 degrees C or 60 degrees C.
-If you are washing at 68 degrees C, inadequate washing is probably not causing your high background. In our experience, raising wash temperatures above 68 degrees C does not decrease background.

compliance-icons
Product Identifier
  • ULTRAhyb Ultrasensitive Hybridization Buffer
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Formamide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.