Learn More
Invitrogen™ Urea
Description
This high-purity molecular biology grade Invitrogen urea is ideal for the preparation of denaturing polyacrylamide gels used in nuclease protection assays. Supplied in one bottle containing 1 kg. Urea is thoroughly tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| For Use With (Application) | Nucleic Acid Gel Electrophoresis & Blotting |
| Packaging Type | Bottle |
| Product Type | Urea |
| Product Line | Ambion |
| Purity | Molecular Biology Grade |
| Quantity | 1000 g |
| Shipping Condition | Room Temperature |
| Form | Powder |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.