missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Solution I, RNA Purification

Product Code. 15671752
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15671752 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15671752 Supplier Invitrogen™ Supplier No. 4305891
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Total RNA Chemistry System component

The protocol is designed for use with the 6100 Nucleic Acid PrepStation and 6700 Automated Nucleic Acid Workstation. This Wash Solution is for use with the Total RNA Chemistry System. Total RNA Chemistry makes possible the rapid isolation of total RNA from various samples, including tissue-cultured cells, whole blood, and peripheral blood mononuclear cells (PBMCs).

DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, RNA Extraction

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Equipment) 6100 Nucleic Acid PrepStation, 6700 Automated Nucleic Acid Workstation
Quantity 1000 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.