missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast RNA (10 mg/mL)

Product Code. 10702487
Change view
Click to view available options
Quantity:
10 x 10 mg
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10702487 10 x 10 mg Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10702487 Supplier Invitrogen™ Supplier No. AM7118
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purified Torulla Invitrogen yeast RNA is suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactions inhibited by exogenous RNA, and it is the most inexpensive source of a high-quality coprecipitant.

Purified Torulla Invitrogen yeast RNA is suitable as a coprecipitant in nucleic acid precipitations. It cannot be used in reactions inhibited by exogenous RNA, and it is the most inexpensive source of a high-quality coprecipitant. This total RNA preparation is supplied in ten 1 mL tubes at a concentration of 10 mg/mL in diethyl pyrocarbonate-treated distilled water. It is typically used at a working concentration of 10 to 20 μg/mL.

What is a Co-precipitant?
Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C.
Concentration 10 mg/mL
For Use With (Application) Carrier for Nucleic Acid Precipitation
Product Type Yeast RNA
Product Line Ambion
Quantity 10 x 10 mg
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.