missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ambrisentan
GREENER_CHOICE

Product Code. 18789748
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18789748 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18789748 Supplier Tocris Bioscience™ Supplier No. 5828/50
  • ONLINE EXCLUSIVE PRICE
    570.00 €
    538.65 € / 50mg
    Save 31.35 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective ETA antagonist

Ambrisentan is a selective endothelin receptor ETA antagonist. Displays approximately 200-fold selectivity for ETA over ETB (Ki values are 1 nM and 195 nM for respectively). Prevents endothelin-induced sudden death in rats.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (αS)-α-[(4,6-Dimethyl-2-pyrimidinyl)oxy]-β-methoxy-β-phenylbenzenepropanoic acid
CAS 177036-94-1
Quantity 50 mg
Molecular Formula C22H22N2O4
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.