missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Antibody Labeling Kits

Product Code. 10106334 Shop All Cytiva Products
Click to view available options
Label or Dye:
Cy2 Reactive Dye
Cy2 mono-Functional Dye
Cy3 Reactive Dye
Cy5 Reactive Dye
Product Type:
Cy2 Ab Labeling Kit
Cy2 MAb Labeling Kit
Cy3 Ab Labeling Kit
Cy5 Ab Labeling Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10106334

Brand: Cytiva PA35000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Convenient way to tag antibodies with CyDye fluorescent dyes

CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.

  • Conjugations are carried out under mild conditions and are complete within 1 hour
  • Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings

Order Info

Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For the conjugation of proteins with Cy5 Reactive Dye
Quantity Each
Product Type Cy5 Ab Labeling Kit
pH Range 7 to 7.3
Molecular Weight (g/mol) 975.15
Solubility Cy5 dye: Soluble in cold water and hot water Coupling buffer: Soluble in cold water and hot water Elution buffer: Soluble in cold water and hot water
Content And Storage 2°C to 8°C, protected from light
Label or Dye Cy5 Reactive Dye
Color Blue
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cy5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.