missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
- Conjugations are carried out under mild conditions and are complete within 1 hour
- Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings
Order Info
Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.
Specifications
Specifications
| For Use With (Application) | For the conjugation of proteins with Cy2 Reactive Dye |
| Quantity | Each |
| Product Type | Cy2 Ab Labeling Kit |
| pH Range | 7 to 7.3 |
| Molecular Weight (g/mol) | 896.95 |
| Solubility | Cy2 dye: Soluble in cold water and hot water Coupling buffer: Soluble in cold water and hot water Elution buffer: Soluble in cold water and hot water |
| Content And Storage | 2°C to 8°C, protected from light |
| Label or Dye | Cy2 Reactive Dye |
| Color | Yellow |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?