Learn More
Descrizione
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
- Low nonspecific binding
- High photostability
- High water solubility
- pH insensitive
Specifica
Specifica
| Description | CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy dye labelled nucleotides exhibit the following properties: |
| pH | 7 |
| Molecular Weight (g/mol) | 765.95 (Monofunction), 949.11 (Bisfunction) |
| Product Type | Cy3-dUTP |
| Reagents | Fluorescent Labeling Reagents |
| Wavelength | 570nm |
| Content And Storage | -15°C to -30°C |
| Quantity | 25 nmol |
| Color | Orange |
| For Use With (Application) | Genomic and Protein research, labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping and gene mapping |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.