missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
- For use in ELISA, immunoprecipitation, and blotting applications.
- Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.
Order Info
Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.
Specifications
Specifications
| For Use With (Application) | For the labelling of biological compounds with Cy5 monofunctional dye |
| Quantity | Each |
| Product Type | Cy5 Mono-Reactive Dye Pack |
| Kit Contents | Five foil packs each containing dried dye to label 1mg of Protein |
| Sufficient For | Labeling 1mg Protein or 30nM Amino groups |
| Molecular Weight (g/mol) | 791.99 |
| Content And Storage | 2°C to 8°C, protected from light |
| Label or Dye | Cy5 mono-Functional Dye |
| Color | Blue |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : cy5 (mono-reactive)
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?