missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)

Product Code. 10479724 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Label or Dye:
Cy3
Cy5
Cy5.5
Product Type:
Cy3 Maleimide
Cy5 Maleimide
Cy5.5 Maleimide
Unit Size:
1mg
25mg
5mg
Product Code. Quantity Label or Dye Product Type unitSize
10479724 1 mg Cy5.5 Cy5.5 Maleimide 1mg
11515085 25 mg Cy5.5 Cy5.5 Maleimide 25mg
11545085 25 mg Cy5 Cy5 Maleimide 25mg
11515095 5 mg Cy3 Cy3 Maleimide 5mg
11555085 5 mg Cy5 Cy5 Maleimide 5mg
11525085 25 mg Cy5.5 Cy5.5 Maleimide 5mg
10637475 1 mg Cy3 Cy3 Maleimide 1mg
10248624 1 mg Cy5 Cy5 Maleimide 1mg
8 options
This item is not returnable. View return policy

Product Code. 10479724

Brand: Cytiva PA15631

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides

The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.

  • Suitable for the labeling of antibodies
  • Example protocols included with each dye pack

Antibody labeling

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications
Quantity 1 mg
Product Type Cy5.5 Maleimide
Kit Contents Foil packs, each containing 1, 5 or 25mg dried dye
pH Range 3 to 10
Molecular Weight (g/mol) 1128.42
Solubility Soluble in cold water and hot water
Content And Storage 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Label or Dye Cy5.5
Color Dark Blue
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.