missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL Plex™ Western Blotting Combination Packs

Product Code. 10585545 Shop All Cytiva Products
Change view
Click to view available options
Formulation:
Cy3, Cy5, Amersham Hybond LFP 0.2 PVDF
Cy3, Cy5, Amersham Protran Premium 0.45 NC
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
10585545 Cy3, Cy5, Amersham Protran Premium 0.45 NC 1 set
10449774 Cy3, Cy5, Amersham Hybond LFP 0.2 PVDF 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10585545 Supplier Cytiva Supplier No. RPN998

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Amersham ECL Plex goat-α-mouse IgG-Cy3, 150μg; Amersham ECL Plex goat-α-rabbit IgG-Cy5, 150μg; Amersham ECL Plex Fluorescent Rainbow Markers, 120μL; 45-001-214: Amersham Protran Premium, 10 x 10cm, 10 sheets or 45-001-215: Hybond LFP 0.2μM PVDF, 20 x 20cm, 3 sheets

Complete system optimized for fluorescent Western blotting

  • Improved formulation of Cy3 conjugates for even lower background

TRUSTED_SUSTAINABILITY

Specifications

Formulation Cy3, Cy5, Amersham Protran Premium 0.45 NC
For Use With (Application) ECL Plex western blotting system
Kit Contents ECL Plex Goat-α-mouse IgG-Cy3 150μg
ECL Plex Goat-α-rabbit IgG-Cy5 150μg
ECL Plex Fluorescent Rainbow Markers 120μL
Amersham Protran Premium 0.45 (10 x 10cm)
Quantity 1 Kit: ECL Plex Goat-α-mouse IgG-Cy3: 150 μg, ECL Plex Goat-α-rabbit IgG-Cy5: 150 μg, ECL Plex Fluorescent Rainbow Markers: 120 μL, Amersham Protran Premium 0.45: (10 x 10 cm)
Storage Requirements Varies by component
Product Type Western Blot Kit
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: cy5 (mono-reactive),sodium azide, nitrocellulose, glycerol,sodium dodecyl sulphate, cy3 (mono-reac
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.