missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Prime Western Blotting Detection Reagent

Product Code. 10308449 Shop All Cytiva Products
Change view
Click to view available options
Sufficient For:
1000cm2 membrane
3000cm2 membrane
Unit Size:
Each
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
10308449 1000cm2 membrane Each
12994780 3000cm2 membrane Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10308449 Supplier Cytiva Supplier No. RPN2232
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Includes

Solution A (luminol solution) and Solution B (peroxide solution)

Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity. Cytiva Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.

  • Stable signal enables multiple exposures and makes reagent suitable for large experimental series
  • Provides convenient handling time between detection and completion of experiment
  • Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging), Amersham Hybond™ P (PVDF) membranes and compatible with Amersham Hyperfilm™, Amersham Hybond- ECL (nitrocellulose) membranes
  • Also compatible with Rainbow™ Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers

 

Education and research

TRUSTED_SUSTAINABILITY

Specifications

Product Type ECL Western Blot Detection Reagent
For Use With (Application) Sensitive Quantitative or Confirmatory Western Blot
Sensitivity ∼1 pg
Sufficient For 1000cm2 membrane
Content And Storage Solution A (Luminol solution, 50mL) and Solution B (Peroxide solution, 50mL), 2°C to 8°C
compliance-icons
Product Identifier
  • ECL Prime'
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: trometamol, ethanediol, imidazole
  • UNKNOWN AQUATIC-Contains 4% of components with unknown hazards to the aquatic environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.