missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ Amersham™ ECL™ Western Blotting Detection Reagents

Product Code. 10754557 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
62.5 mL
125 mL
250 mL
Sufficient For:
1000 cm2 Membrane
2000 cm2 Membrane
4000 cm2 Membrane
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Sufficient For Unit Size
10754557 62.5 mL 1000 cm2 Membrane Each
10340125 125 mL 2000 cm2 Membrane Each
10155854 250 mL 4000 cm2 Membrane Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10754557 Supplier Cytiva™ Supplier No. RPN2109
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Use for all routine Western blotting. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world's first commercially available chemiluminescent detection reagents and still one of the most widely used.

  • Detectable on Hyperfilm™ ECL or with CCD imager
  • Uses enhanced luminol-based detection which is suitable for all routine Western Blotting

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Product Type ECL Western Blot Detection Reagent
For Use With (Application) Western Blot
Quantity 62.5 mL
Sensitivity ∼10 pg
Sufficient For 1000 cm2 Membrane
Content And Storage 62.5mL detection reagent, 162.5mL detection reagent 2. 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.