missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Hybond™ PVDF Membranes: Rolls

Product Code. 15239814 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (LxW):
4 m x 254 mm
4 m x 260 mm
Grade:
Hybond LFP
Hybond P
Hybond SEQ
Unit Size:
1 roll
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) Grade Unit Size
15239814 4 m x 260 mm Hybond P 1 roll
15249814 4 m x 260 mm Hybond SEQ 1 roll
15279884 4 m x 254 mm Hybond LFP 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15239814 Supplier Cytiva Supplier No. 10600021
  • ONLINE EXCLUSIVE PRICE
    478.00 €
    403.27 € / 1 rolls
    Save 74.73 €
    15% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.

Supplied in rolls.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2μm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    TRUSTED_SUSTAINABILITY

    Specifications

    Length (Metric) 4 m
    Width (Metric) 260 mm
    For Use With (Application) Compatible with a range of solvents used for rapid destaining
    Binding Capacity 363 to 430 μg IgG/cm2 (Incubation Method), 167 to 199 μg BSA/cm2 (Incubation Method)
    Format Roll
    Material PVDF
    Pore Size 0.2 μm
    Dimensions (LxW) 4 m x 260 mm
    Product Line Amersham Hybond
    Grade Hybond P
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.