missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Use this versatile nitrocellulose membrane for excellent binding affinity for small proteins and peptides, as well as nucleic acids. Cytiva Amersham™ Protran™ NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Packed in rolls.
- 100% pure nitrocellulose
- High affinity for proteins
- Blocking ability
- Compatible with a range of detection methods, including chemiluminescent, chromogenic, and fluorescent
Specifications
Specifications
| Length (Metric) | 4 m |
| Width (Metric) | 20 cm |
| For Use With (Application) | Western blots, dot-blot assays, and other protein or nucleic acid methods |
| Format | Roll |
| Material | 100% Pure Nitrocellulose |
| Pore Size | 0.45 μm |
Safety and Handling
Signal Word
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : nitrocellulose
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?